Would you like to log in again?
PIG® Puncture Repair Kit, New Pig
Supplier: New Pig
PTY203
46000-766EA 219.3 USD InStock
46000-766
PIG® Puncture Repair Kit, New Pig
Spill Control Materials
Contains field-proven tools to stop nearly any drum leak.
  • Includes wooden wedges, pegs, cones, and rubber mallet for fixing larger holes
  • Fix smaller holes with PIG® Repair Putty, washers, screws, lead wool, and tape
A great addition to spill response supplies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty