Would you like to log in again?
Spill-X® Spill Treatment Kit, Acids, New Pig
Supplier: New Pig
GEN528
89205-852CS 316.39 USD InStock
89205-852
Spill-X® Spill Treatment Kit, Acids, New Pig
Spill Control Materials
Packed with spill control agents that neutralize and solidify acid spills
Works on most mineral and organic acids for safe and easy cleanup, including sulfuric, hydrochloric, nitric, and hydrofluoric. Granular base lets you use only what you need; reduces product waste and disposal costs. Speeds spill cleanup to limit worker exposure and minimize production downtime. Non-toxic, free-flowing dry chemical is environmentally safe. Agents packed in easy-to-use shaker bottles. Excellent as refill for Spill-X Portable Spill Treatment Kit
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty