Would you like to log in again?
PIG® Oil-Only Spill Kit in Storage Chest, New Pig
Supplier: New Pig
KIT434
56620-520EA 1219.75 USD InStock
56620-520
PIG® Oil-Only Spill Kit in Storage Chest, New Pig
Spill Control Materials
Weather-resistant, compartmentalized chest is rugged enough to stand on
Prepacked chest provides easy access to contents. Large, color-coded label makes identification fast and accurate. Temporary disposal bags help make cleanup easier.
No wheels. Not UN rated.
PIG Oil-Only Socks and Booms stop spreading spills; PIG Stat-Mat Pads absorb quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty