Would you like to log in again?
Ace-Thred Filter Adapter with Side Outlet, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
5861-07 5861-03
89235-618EA 594.51 USD InStock
89235-618 89235-616
Ace-Thred Filter Adapter with Side Outlet, PTFE, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This bottom adapter is designed for use with reactor bodies and a PTFE reactor.
Fabricated from PTFE, with 3/8" NPT side discharge port. Adapters have a CAPFE O-Ring to seal adapter in Ace-Thred. The #50 size is supplied with a 100 µm polyethylene support disc. It can also be replaced with a screen support using a retainer ring.
Delivery information: The #80 size is supplied with dual filter supports; a 100 µm polyethylene filter and/or a perforated support plate with retainer ring for use with a 350 µm polypropylene screen.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty