Would you like to log in again?
Peptide Vessel with Threaded Side Port and Removable Filter, Ace Glass Incorporated
Supplier: Ace Glass
6404-36 6404-40 6404-44 6404-05 6404-15 6404-02 6404-30 6404-08 6404-11 6404-33
89054-206EA 559.46 USD InStock
89054-206 89054-208 89054-192 89054-194 89054-196 89054-198 89054-200 89054-210 89054-202 89054-204
Peptide Vessel with Threaded Side Port and Removable Filter, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Cylindrical vessel is equipped with a 2mm bore PTFE stopcock at top, angled GPI threaded side port, and #15 Ace-Thred at bottom that accepts a PTFE adapter with removable filter disc and a PTFE shut-off valve
PTFE adapter has a "front-type" seal using a CAPFE (PTFE encapsulated silicone) O-Ring to make a compression seal in the #15 Ace-Thred on vessel. Porosity C (25–50µm) glass filter disc is press fit into recess at top of the adapter and it can be replaced easily. Other end of adapter features a ¼" FNPT thread that allows connection of a 2.5mm bore PTFE shut-off valve with a luer extension. Supplied with a solid cap for side port. Complete item consists of vessel, PTFE adapter, CAPFE O-Ring, and valve.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty