Would you like to log in again?
Amersham CyDye Antibody Labeling Kits, Cytiva
Supplier: Cytiva
Amersham™ CyDye™
PA33000 PA33000 PA35000 PA35000
95017-392EA 660.6 USD InStock
CA95017-388L 95017-392 CA95017-392L 95017-388
Amersham CyDye Antibody Labeling Kits, Cytiva
Protein Linking and Labeling Systems
CyDye™ fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
  • Convenient way to tag antibodies with CyDye fluorescent dyes
  • Antibody kits contain all reagents required for 2×1 mg reactions
Dyes are packaged in premeasured amounts and foil-sealed to ensure consistent labelings. CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
Conjugations are carried out under mild conditions and are complete within 1 h.
Ordering information: Components include two foil packs each containing dried dye to label 1mg of protein; Coupling buffer (2 vials) [1M sodium carbonate buffer, pH 9.3]; Gel filtration column for the separation of labelled protein from non-conjugated dye; Elution buffer (50 ml) for the elution of labelled protein from the column (Phosphate buffered saline, pH 7.2, containing 0.1% sodium azide); safety data sheet; sample collection tubes (4) and transfer pipettes (6).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty