Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thiourea

Supplier: Cytiva
Warning
Synonyms: ThiocarbamideSulfoureaSulphourea

RPN6301 RPN6301
95056-462EA 128.46 USD InStock
CA95056-462L 95056-462
Thiourea
Thiourea
Thiourea is a chaotrope used together with urea in the IPG strip rehydration to solubilise and denature proteins, and improve the solubility of membrane proteins.


Formula: NH₂CSNH₂
MW: 76.12 g/mol
Boiling Pt: (dec.)
Melting Pt: 176…178 °C
Density: 1.435 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9,III
Merck Index: 12,09505
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/67/2297267.jpg
Order Now
Specification Test Results
Application Used together with urea in the IPG strip rehydration solution to solubilize and denature proteins
Color White
Melting/Freezing Point 176-178°C
Molecular Weight (Mr) 76.13
pH Range 5-7
Physical State Solid
Solubility Easily soluble in methanol, soluble in cold and hot water, partially soluble in diethyl ether
Synonym Sulfourea, Thiocarbamide
Wetted Part No

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty