Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodoacetamide, Proteomics Grade

Supplier: Cytiva
Warning
Synonyms: 2-Iodoacetamide

RPN6302 RPN6302
95056-464EA 196.25 USD InStock
95056-464 CA95056-464L
Iodoacetamide, Proteomics Grade
Iodoacetamide
Used for the efficient alkylation of thiols while minimizing oxidation of competing thiol pairs in protein samples.

  • Proteomic grade
  • Easily soluble in cold and hot water

Ultrapure, proteomic-grade iodoacetamide is used for efficient alkylation of thiols while minimizing oxidation of the competing thiol pairs in protein samples.


Formula: ICH₂CONH₂
MW: 184.96 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 93…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty