Would you like to log in again?
High Purity Solution ddNTPs, Cytiva
Supplier: Cytiva
27-2071-01 27-2061-01 27-2045-01 27-2061-01 27-2045-01 27-2081-01 27-2051-01 27-2051-01 27-2081-01 27-2071-01
95038-192EA 340.13 USD InStock
CA95038-200L 95038-192 95038-190 CA95038-204L 95038-204 95038-200 CA95038-196L 95038-196 CA95038-190L CA95038-192L
High Purity Solution ddNTPs, Cytiva
Nucleic Acid Reagents Nucleotides
Easy to use and time saving solutions for applications requiring ddNTPs of high purity, such as dideoxy sequencing and genotyping reactions.
Tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase DNA Polymerase.
The ddNTP set contains 2',3'-Dideoxyadenosine 5'-Triphosphate, 100 mM Solution (ddATP); 2',3'-Dideoxycytidine 5'-Triphosphate, 100 mM Solution (ddCTP); 2',3'-Dideoxyguanosine 5'-Triphosphate, 100 mM Solution (ddGTP); 2',3'-Dideoxythymidine 5'-Triphosphate, 100 mM Solution (ddTTP)
Ordering information: Supplied in purified water as 5 and 100 mM solutions. Available as individual products or in a convenient set.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty