Would you like to log in again?
GFX 96 PCR Purification Kit, Cytiva
Supplier: Cytiva
29489378 28-9034-45 28-9034-45
95026-726EA 2170.88 USD InStock
95026-726 CA95026-726L MPCA29489378
GFX 96 PCR Purification Kit, Cytiva
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The illustra™ GFX™ 96 Purification Kit utilises glass fibre matrix technology in a 96-well format for highly efficient purification of PCR products. Fragments from PCR are captured by the matrix in the presence of a chaotropic salt, and contaminants are conveniently removed by washing the matrix with a buffered ethanol solution.
  • For the purification of up to 96 PCR products (0,1 to 10 kb) simultaneously in as little as 15 minutes
  • High yields of pure DNA recovered in a small volume of water or a low-ionic strength buffer
  • Typical recoveries are >85% for PCR products 100 bp to 10 kb in length; salt removal typically >99%
  • Avoids ethanol precipitations and hazardous organic extractions
Purified DNA is ready for use in most applications, including fluorescent sequencing, microarrays, labelling, hybridisation, ligation, and transformation.
Up to 95% yield when purifying a 910 bp fragment at a starting concentration of 8.4 ng/µl.
Packaging: Kit contains sufficient quantities of binding plates (10), wash plates (10), collection plates (10), capture buffer (blue, 288 ml), wash buffer (yellow, 80 ml), a choice of two elution buffers each 48 ml (Tris-HCl and sterile water) and an instruction booklet.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty