Would you like to log in again?
Disposable Columns Prepacked with Sepharose™, GST GraviTrap™, Cytiva
Supplier: Cytiva
GraviTrap™
28-9523-60 28952360
71002-624EA 941.69 USD InStock
CA71002-624 71002-624
Disposable Columns Prepacked with Sepharose™, GST GraviTrap™, Cytiva
Chromatography Columns
GST GraviTrap contains 10 disposable columns prepacked with 2 ml of Glutathione Sepharose 4B, sufficient for purification of up to 20 mg of fusion protein.
  • Convenient, prepacked gravity-flow columns
  • Allows direct purification of both clarified and unclarified cell lysates
  • Yields up to 20 mg of pure proteins per column
GST GraviTrap contains 10 disposable columns prepacked with 2 mL of Glutathione Sepharose 4B, sufficient for purification of up to 20 mg of fusion protein. GST-tagged proteins are recovered under mild elution conditions (10 mM glutathione), which preserves functionality of the proteins.
The binding capacity of prepacked GST GraviTrap columns, which can be greater than 10 mg protein/mL medium, varies with the nature of the tagged protein and the exact binding conditions used. For increased convenience, GST Buffer Kit contains stock solutions of the buffers needed for purification of GST-tagged proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty