Would you like to log in again?
rProtein A Protein G GraviTrap™, Cytiva
Supplier: Cytiva
GraviTrap™
28985256 28-9852-56
97060-906EA 1246.06 USD InStock
CA28985256 97060-906
rProtein A Protein G GraviTrap™, Cytiva
Chromatography Columns
rProtein A Protein G GraviTrap contains ten columns with 1 ml 50% rProtein A Sepharose Fast Flow and 50% Protein G Sepharose 4 Fast Flow resins.
  • For general-purpose purification of monoclonal and polyclonal antibodies
  • Suitable when the affinity for protein A and protein G is not known
  • GraviTrap prepacked gravity flow columns enable simple one-step purification of proteins in 30 min without the need for equipment
  • High purity: >95% in one step
  • High protein recovery (typically 70 to 80%) due to high binding capacity of ~35 mg IgG/ml resin
This column can be used for manual purification of polyclonal and monoclonal antibodies from cell culture supernatant and biological fluids. It is particularly suitable when the affinity for protein A and protein G is not known. The gravity flow column relies on gravity and is reusable up to five times, so no additional instruments are needed for this protocol.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty