Would you like to log in again?
Ascorbic Acid Test Method: Colorimetric With Test Strips 50 - 100 - 200 - 300 - 500 - 700 - 1000 - 2000 mg/L MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10023.0001 1.10023.0001
EM1.10023.0001CS 12066.13 USD InStock
EM1.10023.0001 CA1.10023.0001
Ascorbic Acid Test Method: Colorimetric With Test Strips 50 - 100 - 200 - 300 - 500 - 700 - 1000 - 2000 mg/L MQuant™, MilliporeSigma
Environmental Test Kits
The ascorbic acid test is suitable for quickly measuring levels of natural ascorbic acid (vitamin C) in foods such as fruit and vegetable juices, soft drinks, and also to check how much ascorbic acid has been added to certain foods as a preservative or antioxidant.
MilliporeSigma # 1.10023.0001 replaces and is identical to MilliporeSigma # 10023-1
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty