Would you like to log in again?
TouchNTuff® 83-500, Sterile Polyisoprene Gloves, Ansell
Supplier: Ansell
112449 384217 384215 112442 384216 384218 384220 384219
89402-730CS 1638.34 USD InStock
89402-730 89402-732 89402-728 89402-738 89402-724 89402-734 89402-726 89402-736
TouchNTuff® 83-500, Sterile Polyisoprene Gloves, Ansell
Gloves
TouchNTuff® single-use thin polymer gloves provide superior tactility and flexible protection for both workers and products in diverse wet and dry environments.
  • Cleanroom compatible
  • Clean, sterile packs
  • Optimal for critical environment applications
Long gloves with beaded cuffs feature a second-skin feel without the risk of allergic reaction from natural rubber latex proteins. Powder-free. Gamma radiation – sterility assurance level (Sal 10⁻⁶).
Certifications: Manufactured in a facility registered to ISO 9001;2008, ISO 13485;2003. Quality control Complies with EN 420:2003; EN 374/1,2,3:2003
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty