Would you like to log in again?
5' Deadenylase, New England Biolabs
Supplier: New England Biolabs
M0331S M0331S
101449-302EA 83 USD InStock
CA101449-302 101449-302
5' Deadenylase, New England Biolabs
Enzymes
Yeast 5´ Deadenylase is a member of the HIT (histidine triad) family proteins and specifically a member of the Hint branch
Mutations in human aprataxin have been known to be involved in the neurological disorder known as ataxia oculomotor apraxia-1. The human protein has been shown to resolve abortive ligation intermediates by removing the AMP at the 5´ end of DNA (AMP-DNA hydrolase activity). It also repairs DNA damage at 3´ ends by removing 3´-phosphate and 3´-phosphoglycolate. Human aprataxin acts on small molecules as well, nucleotide polyphosphates such as diadenosine tetraphosphate (AppppA) as well as lysyl-AMP. The 5´ Deadenylase is encoded by HNT3 gene of S. cerevisiae. NEB has shown this protein is capable of deadenylation from 5´ end of DNA and RNA, leaving the phosphate at 5´ end. It also cleaves AppppA into ATP and AMP. Its activity on lysyl-AMP is not detectable.
It is the yeast orthologue of aprataxin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty