Would you like to log in again?
Alpha 1-2,3 Mannosidase
Supplier: New England Biolabs
P0729S P0729S
101227-746EA 153 USD InStock
101227-746 CA101227-746
Alpha 1-2,3 Mannosidase
Enzymes
α1-2,3 Mannosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of α1-2 and α1-3 linked D-Mannopyranosyl residues from oligosaccharides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty