Would you like to log in again?
Alpha-N-Acetyl-galactosaminidase
Supplier: New England Biolabs
P0734S P0734S
101227-750EA 160 USD InStock
101227-750 CA101227-750
Alpha-N-Acetyl-galactosaminidase
Enzymes
α-N-Acetyl-galactosaminidase is a highly specific exoglycosidase that catalyzes the hydrolysis of α-linked D-N-acetylgalactosamine residues from oligosaccharides and N-glycans attached to proteins.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty