Would you like to log in again?
Bacteroides Heparinase I
Supplier: New England Biolabs
P0735L P0735L P0735S P0735S
102500-014EA 360 USD InStock
102500-014 CA102500-014 102500-012 CA102500-012
Bacteroides Heparinase I
Enzymes
Bacteroides Heparinase I cloned from Bacteroides eggerthii, also called Heparin Lyase I, is active on heparin and the highly sulfated domains of heparan sulfate.
The reaction yields oligosaccharide products containing unsaturated uronic acids which can be detected by UV spectroscopy at 232 nm. Bacteroides Heparinase I cleaves the glycosidic bond between N-sulfated hexosamines and 2-O-sulfated iduronic acid residues as well as the 2-O-sulfated glucuronic acid residues. The 2-O-sulfated uronic acid residue is essential for the activity of Bacteroides Heparinase I and 6-O-sulfation of GlcNS does not hinder enzyme activity. Bacteroides Heparinase I cleaves 2-O sulfated iduronic acid and 2-O sulfated glucuronic acid residues at similar rates. Limited digest of porcine mucosal heparin with the Bacteroides Heparinase I results in heparin oligosaccharides with a lower extent of sulfation as reported.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty