Would you like to log in again?
beta-N-Acetyl-Hexosaminidase F
Supplier: New England Biolabs
P0721S P0721S
101228-550EA 79 USD InStock
CA101228-550 101228-550
beta-N-Acetyl-Hexosaminidase F
Enzymes
β-N-Acetylhexosaminidasef is a recombinant protein fusion of β-N-Acetyl-hexosaminidase and maltose binding protein
It has identical activity to β-N-Acetyl-hexosaminidase. β-N-Acetyl-hexosaminidasef catalyzes the hydrolysis of terminal β-D-N-acetyl-galactosamine and glucosamine residues from oligosaccharides. β-N-Acetyl-hexosaminidasef can be used to remove O-GlcNAc (N-Acetylglucosaminidase on serine/threonine).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty