Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Nicotinamide adenine dinucleotid, oxidized form (NAD, oxidised form)

Supplier: New England Biolabs (NEB)
Synonyms: Nadide Nicotinamide adenine dinucleotide, oxidized NAD+

B9007S B9007S
101417-884EA 37 USD InStock
101417-884 CA101417-884
β-Nicotinamide adenine dinucleotid, oxidized form (NAD, oxidised form)
β-Nicotinamide adenine dinucleotide (NAD, oxidized form)
β-Nicotinamide adenine dinucleotide (NAD+) is included with the PreCR® Repair Mix product

NAD+ is supplied as an aqueous 50 mM solution.


Formula: C₂₁H₂₇N₇O₁₄P₂
MW: 663.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036253
CAS Number: 53-84-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty