Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BG-GLA-NHS, New England Biolabs
Supplier: New England Biolabs
S9151S S9151S
101447-756EA 474 USD InStock
CA101447-756 101447-756
BG-GLA-NHS, New England Biolabs
Protein Linking and Labeling Systems
BG-GLA-NHS is an amine-reactive building block for the one-step synthesis of SNAP-tag® substrates from amine-containing precursors including proteins, peptides or oligonucleotides
This building block allows you to make custom SNAP-tag substrates for labeling SNAP-tag fusion proteins.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty