Would you like to log in again?
CLIP-SURFACE® 547, New England Biolabs
Supplier: New England Biolabs
S9233S S9233S
101447-730EA 396 USD InStock
CA101447-730 101447-730
CLIP-SURFACE® 547, New England Biolabs
Protein Linking and Labeling Systems
CLIP-Surface™ 547 is a photostable red fluorescent substrate that can be used to label CLIP-tag™ fusion proteins on the surface of living cells or in vitro
This cell impermeable substrate (BC-547) is based on the Dyomics dye DY-547 and is suitable for use with standard TAMRA or Cy3 filter sets. It has an excitation maximum at 554 nm and emission maximum at 568 nm. This package includes 50 nmol of CLIP-Surface 547 substrate, sufficient to make 10 ml of a 5 µM CLIP-tag fusion protein labeling solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty