Would you like to log in again?
EcoRI Methyltransferase
Supplier: New England Biolabs
M0211S M0211S
101228-216EA 77 USD InStock
101228-216 CA101228-216
EcoRI Methyltransferase
Enzymes
EcoRI Methyltransferase modifies the internal adenine residue (N6) of the sequence GAATTC.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty