Would you like to log in again?
EPIMARK® Hot Start Taq DNA Polymerase, New England Biolabs
Supplier: New England Biolabs
EPIMARK®
M0490S M0490L
101710-282EA 300 USD InStock
101710-282 101710-284
EPIMARK® Hot Start Taq DNA Polymerase, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
EpiMark Hot Start Taq DNA Polymerase an excellent choice for use on bisulfite-converted DNA
It is a mixture of Taq DNA Polymerase and a temperature-sensitive, aptamer-based inhibitor. The inhibitor binds reversibly to the enzyme, inhibiting polymerase activity at temperatures below 45°C, but releases the enzyme during normal PCR cycling conditions. This permits room temperature reaction assembly with no separate high-temperature incubation step to activate the enzyme. EpiMark Hot Start Taq DNA Polymerase is supplied with 5X EpiMark Hot Start Taq Reaction Buffer, providing robust amplification for bisulfite-converted DNA and AT-rich amplicons.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty