Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Exonuclease V (RecBCD)
Supplier: New England Biolabs
M0345S M0345L
101710-292EA 84 USD InStock
101710-292 101710-290
Exonuclease V (RecBCD)
Enzymes
Exonuclease V, a RecBCD complex from E.coli has several different enzyme activities, including an ATP-dependent single-stranded DNA endonuclease activity, ss- and ds- DNA exonuclease activity.
The hydrolysis in each case is bi-directional (from both the 3´ and 5´ ends) and processive, producing oligonucleotides. All Exonuclease V activities have divalent cation requirements. Mg2+ is required for the exonuclease activity, while Ca2+ inhibits the exonuclease activity and allows double-stranded DNA unwinding (helicase activity) without hydrolysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty