Would you like to log in again?
M-MuLV Reverse Transcriptase
Supplier: New England Biolabs
M0253S M0253L M0253L M0253S
101228-286EA 348 USD InStock
CA101228-286 101228-286 CA101228-288 101228-288
M-MuLV Reverse Transcriptase
Enzymes
Moloney Murine Leukemia Virus (M-MuLV, MMLV) Reverse Transcriptase is an RNA-directed DNA polymerase
This enzyme can synthesize a complementary DNA strand initiating from a primer using either RNA (cDNA synthesis) or single-stranded DNA as a template. M-MuLV Reverse Transcriptase lacks 3´ → 5´ exonuclease activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty