Would you like to log in again?
Nt.BsmAI
Supplier: New England Biolabs
R0121S R0121S
101449-350EA 80 USD InStock
CA101449-350 101449-350
Nt.BsmAI
Nucleic Acid Reagents Restriction Enzymes
An E.coli strain expressing an altered form of the BsmAI restriction genes from Bacillus stearothermophilus (A664).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty