Would you like to log in again?
ONETAQ® RT-PCR Kit, New England Biolabs
Supplier: New England Biolabs
ONETAQ®
E5310S
101710-294EA 169 USD InStock
101710-294
ONETAQ® RT-PCR Kit, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
OneTaq® RT-PCR Kit combines two powerful mixes, M-MuLV Enzyme Mix and OneTaq Hot Start 2X Master Mix with Standard Buffer for 2-step RT-PCR applications.
The two mixes require minimal handling during reaction setup and yet offer consistent and robust RT-PCR reactions. The first strand cDNA synthesis is achieved by using two optimized mixes, M-MuLV Enzyme Mix and M-MuLV Reaction Mix. M-MuLV Enzyme Mix combines M-MuLV Reverse Transcriptase and murine RNase Inhibitor while M-MuLV Reaction Mix contains dNTPs and an optimized buffer. The kit also contains two optimized primers for reverse transcription and nuclease-free water. An anchored oligo-dT primer [d(T)23VN] forces the primer to anneal to the beginning of the polyA tail. The optimized Random Primer Mix provides random and consistent priming sites covering the entire RNA templates including both mRNAs and non-polyadenylated RNAs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty