Would you like to log in again?
Poly (U) Polymerase , New England Biolabs
Supplier: New England Biolabs
M0337S M0337S
101449-308EA 105 USD InStock
101449-308 CA101449-308
Poly (U) Polymerase , New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
Poly(U) Polymerase catalyzes the template independent addition of UMP from UTP or AMP from ATP to the 3´ end of RNA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty