Would you like to log in again?
Protein A Magnetic Beads, New England Biolabs
Supplier: New England Biolabs
S1425S S1425S
101227-356EA 208 USD InStock
CA101227-356 101227-356
Protein A Magnetic Beads, New England Biolabs
Beads Magnetic Beads
Protein A Magnetic Beads are an affinity matrix for the small-scale isolation and purification of immunoglobulins.
A truncated form of recombinant Protein A is covalently coupled to a nonporous paramagnetic particle. Protein A exhibits high affinity for subclasses of IgG from many species including human, rabbit and mouse. The protein is coupled through a linkage that is stable and leak resistant over a wide pH range. This permits the immunomagnetic purification of IgGs from ascites, serum or cell culture supernatants; the matrix can then be regenerated without loss of binding capacity. Protein A Magnetic Beads can be used to immunoprecipitate target proteins from crude cell lysates using selected primary antibody. In addition, specific antibodies can be chemically cross-linked to the Protein A coated surface to create a reusable immunoprecipitation bead, avoiding the co-elution of antibody with target antigen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty