Would you like to log in again?
pTWIN1 Vector DNA, New England Biolabs
Supplier: New England Biolabs
N6951S
101227-168EA 146 USD InStock
101227-168
pTWIN1 Vector DNA, New England Biolabs
Vectors, Plasmids and Libraries
pTWIN1 is an E.coli expression vector which can be used with the IMPACT™ Kit (NEB #E6901).
pTWIN vectors are designed for protein purification or for the isolation of proteins with an N-terminal cysteine and/or a C-terminal thioester (1). A polylinker in the vector is designed for the in-frame fusion of a target gene between the modified Ssp DnaB (2) and Mxe GyrA inteins. The presence of the chitin binding domain from Bacillus circulans facilitates purification. The double-stranded vector is 7,375 base pairs in length.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty