Would you like to log in again?
pTYB21 Vector, New England Biolabs
Supplier: New England Biolabs
N6709S N6709S
101641-054EA 153 USD InStock
CA101641-054 101641-054
pTYB21 Vector, New England Biolabs
Vectors, Plasmids and Libraries
pTYB21 is an E
It is an N-terminal fusion vector designed for in-frame insertion of a target gene into the polylinker, downstream of the intein tag (the Sce VMA intein/chitin binding domain, 55 kDa). This allows the N-terminus of the target protein to be fused to the intein tag. The self-cleavage activity of the intein allows the release of the target protein from the chitin-bound intein tag, resulting in a single column purification of the target protein.
coli cloning and expression vector (7514 bp) used in the IMPACT™ Protein Purification System which allows the overexpression of a target protein as a fusion to a self-cleavable affinity tag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty