Would you like to log in again?
RNA 5’ Pyrophosphohydrolase
Supplier: New England Biolabs
M0356S M0356S
101641-568EA 108 USD InStock
CA101641-568 101641-568
RNA 5’ Pyrophosphohydrolase
Enzymes
The bacterial RNA 5´ Pyrophosphohydrolase (RppH) removes pyrophosphate from the 5´ end of triphosphorylated RNA to leave a 5´ monophosphate RNA
The RppH protein was also known as NudH/YgdP which can split diadenosine penta-phosphate to ADP and ATP.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty