Would you like to log in again?
RNase B
Supplier: New England Biolabs
P7817S P7817S
101228-572EA 79 USD InStock
CA101228-572 101228-572
RNase B
Enzymes
RNase B is a high mannose glycoprotein that can be used as a positive control for endoglycosidases that cleave N-linked carbohydrates
RNase B has a single N-linked glycosylation site which makes it ideal for SDS-PAGE gel shift assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty