Would you like to log in again?
SNAP-CELL® Block , New England Biolabs
Supplier: New England Biolabs
S9106S S9106S
101447-662EA 164 USD InStock
CA101447-662 101447-662
SNAP-CELL® Block , New England Biolabs
Protein Linking and Labeling Systems
SNAP-Cell® Block (bromothenylpteridine, BTP) is a non-fluorescent compound that blocks the reactivity of the SNAP-tag® in solution or in living cells
It can be used to generate inactive controls in live cell labeling experiments performed with SNAP-tag fusion proteins. SNAP-Cell Block is highly membrane permeable and once in the cell reacts with the SNAP-tag, irreversibly inactivating it for subsequent labeling steps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty