Would you like to log in again?
SNAP-SURFACE® Alexa Fluor 488 , New England Biolabs
Supplier: New England Biolabs
S9129S S9129S
101447-676EA 440 USD InStock
CA101447-676 101447-676
SNAP-SURFACE® Alexa Fluor 488 , New England Biolabs
Protein Linking and Labeling Systems
SNAP-Surface® Alexa Fluor® 488 is a photostable fluorescent substrate that can be used to label SNAP-tag® fusion proteins in solution or on the surface of living cells
This cell impermeable substrate is based on BG-Alexa Fluor 488 and is suitable for standard fluorescein filter sets. It has an excitation maximum at 496 nm and emission maximum at 520 nm. This package includes 50 nmol of SNAP-Surface Alexa Fluor 488 substrate, sufficient to make 10 ml of a 5 µM SNAP-tag fusion protein labeling solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty