Would you like to log in again?
SNAP-SURFACE® Alexa Fluor 647 , New England Biolabs
Supplier: New England Biolabs
S9136S S9136S
101447-686EA 440 USD InStock
CA101447-686 101447-686
SNAP-SURFACE® Alexa Fluor 647 , New England Biolabs
Protein Linking and Labeling Systems
SNAP-Surface Alexa Fluor 647 is a photostable fluorescent substrate that can be used to label SNAP-tag® fusion proteins in solution or on the surface of living cells
This cell impermeable substrate is based on Alexa Fluor 647 and is suitable for standard 635 nm or 650 nm diode laser excitation. It has an excitation maximum at 652 nm and emission maximum at 670 nm. This package includes 50 nmol of SNAP-Surface Alexa Fluor 647 substrate, sufficient to make 10 ml of a 5 µM SNAP-tag fusion protein labeling solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty