Would you like to log in again?
SP6 RNA Polymerase, New England Biolabs
Supplier: New England Biolabs
M0207S M0207S
101228-202EA 78 USD InStock
CA101228-202 101228-202
SP6 RNA Polymerase, New England Biolabs
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
Bacteriophage SP6 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the SP6 phage promoter
The 98.5 KD polymerase catalyzes in vitro RNA synthesis from a cloned DNA template under the SP6 promoter. RNA synthesized using the SP6 RNA Polymerase is suitable for many applications in research and biotechnology.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty