Would you like to log in again?
Streptavidin, New England Biolabs
Supplier: New England Biolabs
N7021S N7021S
101228-520EA 85 USD InStock
CA101228-520 101228-520
Streptavidin, New England Biolabs
Protein Linking and Labeling Systems
A 52.8 kDa non-glycosylated protein with a very high affinity for biotin
Due to its high affinity for biotin, it is used to bridge biotinylated probes and biotinylated enzymes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty