Would you like to log in again?
T4 Gene 32
Supplier: New England Biolabs
M0300L M0300S M0300S M0300L
101228-376EA 360 USD InStock
CA101228-376 101228-376 CA101228-378 101228-378
T4 Gene 32
Proteins and Peptides
T4 Gene 32 Protein is a single-stranded DNA (ssDNA) binding protein required for bacteriophage T4 replication and repair. It cooperatively binds to and stablizes transiently formed regions of ssDNA and plays an important structural role during T4 phage replication. It also has been used extensively to stabilize and mark regions of ssDNA for electron microscopic examination of intracellular DNA structures. Recently, it has been shown to improve restriction enzyme digestion, improve the yield and efficiency of reverse transcription (RT) reactions during RT-PCR, enhance T4 DNA polymerase activity, as well as increase the yield of PCR products.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty