Would you like to log in again?
E. coli Topoisomerase I
Supplier: New England Biolabs
M0301S M0301L M0301S M0301L
101227-646EA 81 USD InStock
101227-646 CA101227-646 101227-644 CA101227-644
E. coli Topoisomerase I
Enzymes
Topoisomerase I (E.coli) catalyzes the relaxation of negatively supercoiled DNA.
Topoisomerase I has also been implicated in knotting and unknotting DNA and in linking complementary rings of single-stranded DNA into double-stranded rings. The intact holoenzyme is a 97 kDa protein.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty