Would you like to log in again?
Tth Endonuclease IV
Supplier: New England Biolabs
M0294S M0294S
101446-504EA 85 USD InStock
CA101446-504 101446-504
Tth Endonuclease IV
Enzymes
Tth Endonuclease IV is a thermostable apurinic/apyrimidinic (AP) endonuclease from Thermus thermophilus
Tth Endo IV will hydrolyze an AP site at the first phosphodiester bond 5' to the lesion leaving a 3' hydroxyl and a deoxyribose 5'-phosphate at the 5' terminus. The enzyme also has a 3'-diesterase activity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty