Would you like to log in again?
Rabbit Complement Type (HLA-DR), MP Biomedicals
Supplier: MP Biomedicals
08642812
IC642812EA 216.91 USD InStock
IC642812
Rabbit Complement Type (HLA-DR), MP Biomedicals
Sera (Serum)
The complement is produced from a large pool of New Zealand White rabbits, 8 - 12 weeks old
Reconstitute the lyophilized serum with an equal amount of sterile distilled water. After reconstitution store at 2 - 8 °C and use within eight hours. Discard any unused thawed complement after eight hours.
The animals are of United States origin. Cytotoxic titer is determined by NIH standards for microlymphocytotoxicity. Minimum release criteria is 80 - 100% cell death in 1:2 dilution.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty