Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Human Serum Goat Polyclonal Antibody

Supplier: MP Biomedicals
08682651
IC682651EA 211.16 USD InStock
IC682651
Anti-Human Serum Goat Polyclonal Antibody
Antibodies
Anti-Human
Host: goat
Isotype: IgG Fraction liquid

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty