Would you like to log in again?
SP Bel-Art Tubing Sizer, Metric Gauge, Bel-Art Products, a part of SP
Supplier: Bel-Art
Scienceware®
F13390-0000
89428-758EA 74.17 USD InStock
89428-758
SP Bel-Art Tubing Sizer, Metric Gauge, Bel-Art Products, a part of SP
Tubing Selectors
This handy metric diameter gauge will end doubt about inside or outside diameters.
  • Polypropylene construction
  • Measures both inside and outside diameters
  • Autoclavable
  • Metric diameter gauge allows easy sizing of tubing
The gauge measures glass and plastic tubing, connecting pieces, rods, test tubes, and more between 1.5 mm and 16 mm. Holes on one side measure O.D., while pins on the other side measure I.D. Order one for your tubing and one for your test tubes, centrifugation tubes, and adaptors. Made of polypropylene, the gauge is autoclavable at 121 °C (250 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty