Would you like to log in again?
Blood Cell Counting Vial, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
107047
89009-318CS 277.41 USD InStock
89009-318
Blood Cell Counting Vial, Globe Scientific
Vials
These cell counter vials come with a polyethylene snap-on lid.
The vials are graduated at the 10 and 20 ml marks.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty