Would you like to log in again?
Corning® Dispase
Supplier: Corning
Corning®
354235 354235
76176-668EA 678.49 USD InStock
76176-668 CACB354235
Corning® Dispase
Enzymes
Dispase is a bacillus-derived neutral metalloprotease that is recommended for recovering cell cultured on Corning® Matrigel® Basement Membrane Matrix. Dispase will yield a single cell suspension far more gently and effectively than trypsin, collagenase, or other proteolytic enzymes; it will not harm cells harvested for sub cultivation or bioassays. In addition, Dispase may be used for tissue dissociation. Dispase cleaves fibronectin, collagen IV, and to a lesser extent collagen I, but is does not cleave collagen V or laminin.
  • Tested for ability to solubilise a 1 mm thick layer of gelled BD Matrigel™ matrix after two hours at 37 °C
  • Filtered and tested and found negative for bacteria, fungi, and mycoplasma
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty