Would you like to log in again?
Corning® Human Fibronectin
Supplier: Corning
Corning®
356008 356008 356009 356009 354008 354008
47743-654EA 467.53 USD InStock
CACB-356009A 47743-654 47743-730 47743-728 CACB354008 CACB356008
Corning® Human Fibronectin
Proteins and Peptides
Fibronectin is found in interstitial matrix and plasma. The principal function of fibronectin appears to be in cellular migration during wound healing and development. It can be used as a thin coating on tissue culture surfaces to promote attachment, spreading and proliferation of a variety of cell types.
Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi and mycoplasma. Source material tested for hepatitis B antigen and HIV-1 antibody.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty