Would you like to log in again?
Synthetic Poly-D-Lysine, Corning®
Supplier: Corning
Corning®
354210 354210
47743-736CS 149.25 USD InStock
CACB354210 47743-736
Synthetic Poly-D-Lysine, Corning®
Proteins and Peptides
Poly-D-lysine (PDL) is a synthetic molecule used as a coating to enhance cell attachment to plastic and glass surfaces. It has been used to culture a wide variety of cell types, particularly neurons, glial cells, and transfected cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty