Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Transport Tube, 50 ml, Globe Scientific

Supplier: Globe Scientific
6255MF
89497-774CS 341.44 USD InStock
89497-774
Transport Tube, 50 ml, Globe Scientific
Tubes Test Tubes
These 50 ml transport tubes are leak-resistant and ideal for centrifugation, transport, storage, and specimen collection.

  • The tube is metal-free
  • Autoclavable (without screw cap)

This conical bottm, self standing tube offers a spin rate of 5000 rpm.

Tubes have a skirted conical bottom and feature white printed graduation marks and a white frosted writing area.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty